Order Entry
ContactUsLinkComponent
Hemoccult* SENSA* Fecal Occult Blood Slide Test System
Hemoccult II® SENSA® Fecal Occult Blood Test Systems, Beckman Coulter®
Catalog #: CA74020-006
Supplier:  Beckman Coulter
CAS Number:  
Hemoccult* SENSA* Fecal Occult Blood Slide Test System
Hemoccult II® SENSA® Fecal Occult Blood Test Systems, Beckman Coulter®
Catalog #: CA74020-006
Supplier:  Beckman Coulter
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Description:
    Single Slides System
  • Cat. no.:
    CA74020-006
  • Supplier No.:
    64151A

Specifications

About this item

Rapid, convenient, and qualitative method features improved readability and higher clinical sensitivity for detecting fecal occult blood.

  • Qualitative
  • Sensitivity 92%
  • Triple-slide

Triple-slide products are designed so patients can collect serial specimens over three days, increasing the probability of detecting hidden blood.

Certifications: CLIA waived.
Ordering information: The Dispensapak™ Plus system is supplied with 40 patient kits and two 15mL bottles of developer. Single Slides pack 10012-018 is supplied with 100 slides, 100 applicator sticks, and two 15mL bottles of developer. Single Slides 10012-020 is packaged as a case with 1000 slides, 1000 applicator sticks, and twenty 15mL bottles of developer.
Customers Who Bought This Also Bought