Sample Tubes, 50 ml, Simport Scientific
Catalog #: CA73960-336
Supplier: Simport Scientific
Specifications
- Capacity:50 ml (1.6 oz.)
- Diameter ×L:30 x 115 mm (11/8 x 41/2")
- Sterility:Sterile
- Cat. no.:CA73960-336
- Cap Color:Green
- Supplier No.:C571-1
Specifications
About this item
These polypropylene, conical bottom tubes with self-standing bases have molded-in graduations from 10 to 50 ml in 5 ml increments.
- Chemical-resistant and shatterproof
- With a leakproof screw cap, which is particularly important for transporting hazardous material
Ideal for transport and storage of urine, sputum, and most liquids or particulate samples.
Temperature: –90 to 121 °C (–130 to 250 °F).
Customers Who Bought This Also Bought