ContactUsLinkComponent
HISTOSETTE* II Tissue Processing/Embedding Cassette
Histosette® II Tissue Processing/Embedding Cassettes with Integrated Lids, Simport
Catalog #: CA73960-060
Supplier:  Simport Scientific
Histosette® II Tissue Processing/Embedding Cassettes with Integrated Lids, Simport
Catalog #: CA73960-060
Supplier:  Simport Scientific
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
  • Color:
    White
  • Cat. no.:
    CA73960-060
  • Supplier No.:
    M492-2
Specifications
About this item
These disposable plastic cassettes hold tissue specimens during processing and embedding, as well as in storage
These disposable plastic cassettes hold tissue specimens during processing and embedding, as well as in storage
Molded from a special high-density polymer, they can keep specimens safely submerged in liquid and are resistant to the chemical action of histological solvents. The efficient flow-through slots maximize fluid exchange and ensure proper drainage. The one-piece integral lids can be opened and closed as often as necessary and will always relock securely without danger of specimen loss. The unique front hinge design allows for single-hand cassette manipulation. The anterior writing area is slanted at a 45° angle to make the cassette more suitable with certain types of cassette labeling instruments
Ordering information: Packed in three dispenser packs of 500 cassettes.
Customers Who Bought This Also Bought