To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Color:Teal
- Description:Swab, liquid Stuart's
- Cat. no.:CA73270-005
- Type:Single rayon plastic shaft
- Supplier No.:SP132
Specifications
About this item
Sterile, ready to use units for the collection and transport of bacteriological specimens
Both Amies provide excellent environments for the survival of anaerobes through the complete immersion of the swabs into the gel and by the presence of the reducing agent sodium thioglycollate. Shelf life is 24 months. Amies Clear is a modified Amies gel that provides excellent survival for a wide range of bacteria and is compatible with gram stain. Amies Charcoal enhances the survival of fastidious organisms by neutralizing any toxins that may be present in the specimen.
Sterile, ready to use units for the collection and transport of bacteriological specimens
Both Amies provide excellent environments for the survival of anaerobes through the complete immersion of the swabs into the gel and by the presence of the reducing agent sodium thioglycollate. Shelf life is 24 months. Amies Clear is a modified Amies gel that provides excellent survival for a wide range of bacteria and is compatible with gram stain. Amies Charcoal enhances the survival of fastidious organisms by neutralizing any toxins that may be present in the specimen.
Both Amies provide excellent environments for the survival of anaerobes through the complete immersion of the swabs into the gel and by the presence of the reducing agent sodium thioglycollate. Shelf life is 24 months. Amies Clear is a modified Amies gel that provides excellent survival for a wide range of bacteria and is compatible with gram stain. Amies Charcoal enhances the survival of fastidious organisms by neutralizing any toxins that may be present in the specimen.
Modified Liquid Stuart's provide excellent recovery in routine bacteriology and are compatible with gram stain and many rapid screening methods. Shelf life is 18 months.
Swab length is 15.2cm (6"). Color-coded cap with protective overskirt. Cap rings ensure containment of biohazardous material. Tube label shows code, lot number, and expiration date. Thick tube wall protects medium from dehydration. Spiral media chamber keeps sample immersed and protected during transport. Round-bottom tube for easy racking. Seamless molding ensures tube strength and integrity. Rayon swab tip is inert, non-toxic, and permits good sample retrieval and absorption.