ContactUsLinkComponent
Diff-Safe* Blood Dispenser
Diff-Safe® Blood Dispensers, Alpha Scientific
Catalog #: CA73000-077
Supplier:  Alpha Scientific
Diff-Safe* Blood Dispenser
Diff-Safe® Blood Dispensers, Alpha Scientific
Catalog #: CA73000-077
Supplier:  Alpha Scientific
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    Blood Dispensers
  • Cat. no.:
    CA73000-077
  • Supplier No.:
    101
Specifications
About this item
Designed to minimize airborne pathogens by eliminating the need to remove the stopper from the blood tube
Designed to minimize airborne pathogens by eliminating the need to remove the stopper from the blood tube
The dispensers feature a cannula that inserts easily through the rubber stopper of a blood collection tube. When the tube is inverted and the dispenser pressed against a slide, it dispenses a drop ideally sized for making a blood smear. Compatible with BD Vacutainer® tubes with BD Hemogard™ closures. Nonsterile.
Customers Who Bought This Also Bought