Order Entry
ContactUsLinkComponent
OSOM* Trichomonas Rapid Test
OSOM® Trichomonas Rapid Test Kit, Sekisui Diagnostics
Catalog #: CA71001-052
CAS Number:  
OSOM* Trichomonas Rapid Test
OSOM® Trichomonas Rapid Test Kit, Sekisui Diagnostics
Catalog #: CA71001-052
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Description:
    Trichomonas Rapid Test Kit
  • Cat. no.:
    CA71001-052
  • Supplier No.:
    181
  • No. of Tests:
    25

Specifications

About this item

The Trichomonas Rapid Test is an immunochromatographic assay designed to detect Trichomonas Vaginalis antigens directly from vaginal swabs, so a speculum may not be required

Control kit (sold separately) includes additional controls for the trichomonas test.

Detects the antigen, removing the uncertainty with live organism instability. Sensitivity is 83% versus culture. Studies show wet mount sensitivity is <60%. Kits should be stored at room temperature to maintain a 16-month shelf life.

Certifications: CLIA-waived. CE marked.
Ordering information: Test kit includes controls. Control kit includes 10 positive control swabs.
Customers Who Bought This Also Bought