Large Plug for DriSolv® Solvents, EMD Millipore
Catalog #: CA692012-2
Supplier: MilliporeSigma
Specifications
- Description:Large Plug
- Cat. no.:CA692012-2
- For Use With:DriSolv®
- Supplier No.:692012-2
Specifications
About this item
Customers Who Bought This Also Bought
Sodium thiosulfate 0.10 N in aqueous solution, VWR Chemicals BDH®
Supplier: VWR International
$153.23 to $602.78
Methanol ≥99.9%, OmniSolv®, gradient grade for HPLC, Supelco®
Supplier: MilliporeSigma
$635.20 to $2,541.50
Water ACS, Reagent Grade ASTM Type I, ASTM Type II Packaged in plastic containers
Supplier: Ricca Chemical
$200.03
Adhesive Labels on Rolls, Round, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
$77.45
Pierce™ Guardian™ Peroxidase Conjugate Stabilizer/Diluent, Thermo Scientific
Supplier: Thermo Scientific
$410.82
Sodium bisulfite ≥90% FCC, purified, Macron Fine Chemicals™
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
$237.49 to $1,424.99