Order Entry
ContactUsLinkComponent
ICON 25 hCG 25 Test
ICON® 25 hCG Pregnancy Test, Beckman Coulter®
Catalog #: CA63900-010
Supplier:  Beckman Coulter
CAS Number:  
ICON 25 hCG 25 Test
ICON® 25 hCG Pregnancy Test, Beckman Coulter®
Catalog #: CA63900-010
Supplier:  Beckman Coulter
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Description:
    ICON® 25 hCG Pregnancy Test
  • Cat. no.:
    CA63900-010
  • Supplier No.:
    43025A

Specifications

About this item

Highly sensitive, one-step immunochemical rapid test for qualitative detection of human chorionic gonadotropin in urine or serum to aid in early pregnancy detection.

  • Sensitive (25 mIU/mL)
  • Specificity > 99%
  • One-step (test time 3 minutes for urine, 5 minutes for serum)
  • Easy to use

Meets the requirements of physicians and hospitals for ease-of-use, value, and accuracy.

Certifications: Urine is CLIA waived; serum is moderately complex.
Ordering information: Kit contents include 25 test devices and droppers, and two additional disposable droppers.
Customers Who Bought This Also Bought