Nunc® Lab-Tek™ II Chamber Slide™ System, Sterile, Thermo Scientific
Catalog # CA62407-297
Supplier: Thermo Fisher Scientific
Specifications
- Cat. no.:CA62407-297
- Working Volume:0.2 - 0.5 ml
- No. of Chambers:8
- Well Area:0.7 cm2
- Supplier No.:154534
Specifications
About this item
This system features removable, polystyrene chambers that are sealed to a 25x75mm non-fluorescent glass slide using biocompatible acrylic adhesive
Numbers along the border of the chamber and a Superfrost® writing area on the slide facilitate identification.
The sealant remains with the media chamber after detachment, leaving a hydrophobic border to define cell culture areas. A tabbed cover fits over the media chamber allowing gas exchange while minimizing evaporation. The slides are provided sterile with an RS wash to create a consistent growth surface for adherent cell lines.
Frequently Bought Together