Nunc® Lab-Tek™ II Chambered Coverglass, Thermo Scientific
Catalog #: CA62407-056
Supplier: Thermo Fisher Scientific
Specifications
- Cat. no.:CA62407-056
- Supplier No.:155409
Specifications
About this item
Culture and view living cells on a single device
Thermo Scientific™ Nunc™ Lab-Tek™ II Chambered Coverglass includes a thin coverglass that provides the optimum optical characteristics required for high-magnification microscopy and confocal image analysis.
Customers Who Bought This Also Bought