Falcon® Cell Culture Inserts, Corning
Catalog #: CA62406-198
Supplier: Corning
Specifications
- Pore size:8.0 µm
- Sterility:Sterile
- Description:Inserts for 24-well plates
- Cat. no.:CA62406-198
- Membrane:Transparent PET membrane
- Supplier No.:353097
Specifications
About this item
Track-etched PET membranes have a smooth surface and defined cylindrical pores that traverse the membrane.
- Low protein binding PET membrane
- Sterilized by gamma irradiation
- A wide variety of configurations including 6, 12 and 24 well
- A broad selection of membrane pore sizes, 0.4, 1.0, 3.0 and 8.0 µm diameter
- Packed in individual blister packs, 48 inserts/case
- Non-Tissue Culture-treated insert housings prevent promiscuous growth of cells on the insert walls
- Innovative hanging design facilitates pipetting and allows for co-culture
Customers Who Bought This Also Bought