Specifications
- Optical property:Transparent
- Sterility:Non sterile
- Description:Sealing film
- Supplier No.:T329-1
Specifications
About this item
Transparent, adhesive sealing film is ideal for reducing cross contamination and/or spill over in PCR applications.
- Thermostable and functional from −70 to +95 °C
- Choice of sterile or non sterile
- Pierceable with a pipette tip for easy access to sample
Aluminium sealing foil is suitable for both manual sealing during PCR work and high throughput applications. It is recommended that the foil be applied using the specially designed rubber roller to ensure a perfect bond, eliminating the risk of evaporation.