Biotube™ Cluster Tube System, Simport
Catalog #: CA60872-494
Supplier: Simport Scientific
Specifications
- Sterility:Sterile
- Color:—
- Description:Plug Caps, Strips of 8
- Cat. no.:CA60872-494
- Supplier No.:T100-35
Specifications
About this item
The Biotube™ Cluster Tube System is a polypropylene tube and rack system with tubes held in a standard 8×12 configuration.
- Alphanumeric coding and removeable 96-place ID card for accurate sample location
- Transparent cover keyed to base to prevent misalignment
- Ideal for storing, freezing and transporting reagents and specimens
- Racks and tubes can be used over a wide temperature range, −80 to 121 °C
Tubes and racks can be autoclaved up to 100 times and will resist most research chemicals. The tube supporting grid plate can be removed and used as a self-standing support. It can also be placed in a refrigerator or freezer for improved air circulation around the tubes, or in water baths to allow controlled warming of the tubes contents.
Customers Who Bought This Also Bought