Order Entry
ContactUsLinkComponent
PLUG CAPS 17MM PE YELLOW PK1000
Plug Type Push-in Caps, Simport Scientific
Catalog #: CA60830-190
Supplier:  Simport Scientific
PLUG CAPS 17MM PE YELLOW PK1000
Plug Type Push-in Caps, Simport Scientific
Catalog #: CA60830-190
Supplier:  Simport Scientific

Specifications

  • Color:
    Yellow
  • Material:
    PE
  • Cat. no.:
    CA60830-190
  • Packaging:
    1000/Bag
  • For Tubes:
    17 mm
  • Supplier No.:
    T401-10Y

Specifications

About this item

Plug-type push-in polyethylene caps are designed for test tubes with an outside diameter of 12 mm. The 17 mm caps are designed for T408 centrifuge tubes.

  • Anaerobic use or for storage, transfer and centrifuge applications
  • Not autoclavable
  • Six colors available

Two size options are available for these plug-type caps.

Customers Who Bought This Also Bought