8110S N95 Particulate Respirator, 3M™
Catalog #: CA56621-960
Supplier: 3M
Specifications
- Description:8110S N95 Particulate Respirator
- Cat. no.:CA56621-960
- Supplier No.:70070757078
Specifications
About this item
Designed for people with smaller faces, this respirator is ideal for grinding, sanding, sweeping, bagging, and other dusty operations
Caution: All 3M™ products must be used in accordance with OSHA regulations and the user instructions, warnings, and limitations accompanying each product.
Features 3M’s Advanced Electret Media for easier and cooler breathing and adjustable noseclip for a secure seal and to reduce eyewear fogging. NIOSH-approved to provide at least 95% filtration efficiency against certain solid and liquid aerosols that do not contain oil.
Ordering information: This is the smaller version of the 3M 8210 N95 Respirator.
Customers Who Bought This Also Bought