8511 N95 Respirator, 3M™
Catalog #: CA56621-936
Supplier: 3M
Specifications
- Description:8511 N95 Respirator
- Cat. no.:CA56621-936
- Supplier No.:70070757557
Specifications
About this item
Respirator provides protection against certain solid and liquid particles that do not contain oil.
- Provides comfortable protection against certain non-oil based particles
- Cool Flow™ exhalation valve reduces heat and humidity buildup
- Features braided headbands, soft inner materials, adjustable "M" nose clip
The innovative “M” nose clip helps provide a custom fit and secure seal against the face. Comfortable, durable respirator has the Cool Flow® exhalation valve to reduce heat and humidity build-up inside the respirator. Provides minimum filtration efficiency of 95%.
Caution: All 3M™ products must be used in accordance with OSHA regulations and the user instructions, warnings, and limitations accompanying each product.
Customers Who Bought This Also Bought