Head and Face Shield Combinations, 3M™
Catalog #: CA56615-707
Supplier: 3M
Specifications
- Color:Clear
- Description:Ratchet Headgear and 3M™ Clear Chin Protector HCP8
- Cat. no.:CA56615-707
- Supplier No.:82521-10000
Specifications
About this item
A combination head, face, and hearing protection solution
- Combination head, face, and hearing protection solution
- Includes six-point ratchet suspension hard hat
- UVicator™ sensor, metal mesh visor, earmuff, rainshield, and visor seal
- Requires protective eyewear (sold separately)
Combinations include a six-point ratchet suspension hard hat with UVicator™ sensor, metal mesh visor, earmuff, rainshield, and visor seal. Requires protective eyewear, which is sold separately.
Certifications: Items meet ANSI Z87.1-2003 standards.
Customers Who Bought This Also Bought