Specifications
- Description:5501 Series Half-Mask Respirators
- Size:Medium
- Cat. no.:CA56223-222
- Supplier No.:550030M
Specifications
About this item
Comfortable half-mask or full facepiece respirators for use with North Safety Products cartridges and filters.
- Reliable and latex-free respirators offer direct cartridge-to-facepiece connection
- 7700 and 7600 series have nonallergenic silicone facepieces
- 5500 and 5400 series have thermoplastic elastomer facepieces
- NIOSH approved when used with North Safety Products cartridges
Interchangeable cartridges and filters provide air purifying protection for a variety of applications.
Half-mask respirators fit snugly while the cradle suspension system alleviates pressure points. Can be worn with safety glasses or goggles. Full facepiece respirators provide eye, face, and respiratory protection and are made of soft, flexible materials for optimum comfort.
The 7700 and 7600 series respirators have facepieces made of nonallergenic silicone; the 5500 and 5400 series respirators feature facepieces constructed of thermoplastic elastomer. Impact-resistant, wraparound polycarbonate lens permits 200° vision field. Full facepiece respirators include an oral/nasal cup. The 7600 series respirators include a speaking diaphragm.
NIOSH approved when used with North Safety Products cartridges and/or filters. Full facepiece lens meets ANSI Z87.1 standard for impact and penetration resistance. The eyeglass frame for the 7600 series respirators is constructed of sturdy metal and attaches to the lens of the facepiece with a suction cup.