8210 Disposable N95 Respirator, 3M™
Catalog #: CA56222-362
Supplier: 3M
Specifications
- Description:8210 N95 Respirator
- Cat. no.:CA56222-362
- Supplier No.:8210
Specifications
About this item
Economical respirator provides reliable worker protection against certain solid and liquid aerosols that do not contain oil.
- Provides reliable worker protection against certain solid and liquid aerosols that do not contain oil
- Provides minimum filtration efficiency of 95%
- Lightweight construction for greater worker comfort
- Adjustable nose clip for custom fit and secure seal
Provides minimum filtration efficiency of 95%. Lightweight construction for greater worker comfort. Adjustable noseclip for custom fit and secure seal.
Caution: All 3M™ products must be used in accordance with OSHA regulations and the user instructions, warnings, and limitations accompanying each product.
Customers Who Bought This Also Bought