Order Entry
ContactUsLinkComponent
VWR* Micro Cover Glasses, Round, No. 1
VWR® Micro Cover Glasses, Round, No. 1
Catalog #: CA48380-046
Supplier:  VWR International
VWR* Micro Cover Glasses, Round, No. 1
VWR® Micro Cover Glasses, Round, No. 1
Catalog #: CA48380-046
Supplier:  VWR International

Specifications

  • Ø:
    18 mm
  • Numbering:
    #1
  • Description:
    Cover glasses, round
  • Cat. no.:
    CA48380-046

Specifications

About this item

These cover glasses offer maximum clarity, superior corrosion resistance, and uniform thickness and size.

  • Circular. No. 1 micro cover glasses have a thickness of 0.13 to 0.17mm

They are free of striae and oily film.

Ordering information: Packaged 1 oz. per plastic box.
Customers Who Bought This Also Bought