Order Entry
ContactUsLinkComponent
VWR* Micro Cover Glasses, Square, No. 2
VWR® Micro Cover Glasses, Square, No. 2
Catalog #: CA48368-040-1
Supplier:  VWR International
VWR* Micro Cover Glasses, Square, No. 2
VWR® Micro Cover Glasses, Square, No. 2
Catalog #: CA48368-040-1
Supplier:  VWR International

Specifications

  • Numbering:
    #2
  • Description:
    Cover glasses, square
  • L×W:
    18×18 mm
  • Cat. no.:
    CA48368-040-1

Specifications

About this item

These cover glasses offer maximum clarity, superior corrosion resistance, and uniform thickness and size.

  • No. 2 thickness is 0.17 to 0.25mm

They are free of striae and oily film.

Ordering information: Packaged 1 oz. per plastic box.
Customers Who Bought This Also Bought