Order Entry
ContactUsLinkComponent
VWR* Micro Cover Glasses, Square, No. 1-1/2
VWR® Micro Cover Glasses, Square, No. 11/2
Catalog #: CA48366-227-1
Supplier:  VWR International
VWR* Micro Cover Glasses, Square, No. 1-1/2
VWR® Micro Cover Glasses, Square, No. 11/2
Catalog #: CA48366-227-1
Supplier:  VWR International

Specifications

  • Numbering:
    #1.5
  • Description:
    Cover glasses, square
  • L×W:
    22×22 mm
  • Cat. no.:
    CA48366-227-1

Specifications

About this item

These cover glasses offer maximum clarity, superior corrosion resistance, and uniform thickness and size.

  • No. 11/2 thickness is 0.16 to 0.19mm

They are free of striae and oily film.

Ordering information: Packaged 1 oz. per plastic box.
Customers Who Bought This Also Bought