Order Entry
ContactUsLinkComponent
VWR* Micro Cover Glasses, Square, No. 1
VWR® Micro Cover Glasses, Square, No.1
Catalog #: CA48366-089-1
Supplier:  VWR International
VWR* Micro Cover Glasses, Square, No. 1
VWR® Micro Cover Glasses, Square, No.1
Catalog #: CA48366-089-1
Supplier:  VWR International

Specifications

  • Numbering:
    #1
  • Description:
    Cover glasses, square
  • L×W:
    25×25 mm
  • Cat. no.:
    CA48366-089-1

Specifications

About this item

VWR® Micro Cover Glasses are square, corrosion-resistant, and uniformly thick and flat.

  • Corrosion-resistant
  • Uniform thickness

No. 1 thickness is 0.13 to 0.17mm.

Ordering information: Packaged 1 oz. per plastic box.
Customers Who Bought This Also Bought