Frosted Fluorescent Antibody Microscope Slides, Epredia™
Catalog #: CA48349-057
Supplier: Erie Scientific
Specifications
- Description:Slides, flourescent; ring diameter: 15 mm
- L×W:75×25 mm (3×1")
- Thickness:0.9 - 1.1 mm
- Cat. no.:CA48349-057
- Supplier No.:2961
Specifications
About this item
Slides feature two etched circles.
- For fluorescent antibody examination
- Features two etched circles
- No coating
- Tab features durable, easy to write and read finish
They are frosted at one end of one side.
Customers Who Bought This Also Bought