Order Entry
ContactUsLinkComponent
PBS 10X PH 7.4 1L PK6
Corning® Phosphate-Buffered Saline (PBS), 10X Concentrate, pH 7,4, Molecular Biology Grade
Catalog #: CA45001-130
Supplier:  Corning
CAS Number:  
PBS 10X PH 7.4 1L PK6
Corning® Phosphate-Buffered Saline (PBS), 10X Concentrate, pH 7,4, Molecular Biology Grade
Catalog #: CA45001-130
Supplier:  Corning
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Description:
    10X Phosphate-Buffered Saline (PBS)
  • Size:
    6 x 1 L
  • Cat. no.:
    CA45001-130
  • Supplier No.:
    46-013-CM

Specifications

About this item

Phosphate Buffered Saline (PBS) is used to maintain a physiological pH and osmolarity, two key factors in tissue culture and molecular biology research

  • Manufactured in a cGMP environment
  • Formulated with high quality WFI water
  • RNase -, DNase-, and Protease-free
  • PBS without calcium and magnesium
  • pH 7.4 ±0.1
Caution: For research use only.
Customers Who Bought This Also Bought