Order Entry
ContactUsLinkComponent
TRYPSIN EDTA (0.25%) 6X100ML
Corning® Trypsin
Catalog #: CA45000-664
Supplier:  Corning
CAS Number:  
TRYPSIN EDTA (0.25%) 6X100ML
Corning® Trypsin
Catalog #: CA45000-664
Supplier:  Corning
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Size:
    100 ml
  • Storage conditions:
    Frozen
  • Shipping conditions:
    Frozen
  • Enzyme name:
    Trypsin
  • Formulation:
    EDTA 1X 0.05 %(0.25 % for 25-053-CI) Tryp sin/0.53 mM EDTA in HBSS with sodium bicarbonate, without calcium and magnesium
  • Cat. no.:
    CA45000-664
  • Supplier No.:
    25-053-CI

Specifications

About this item

0,05% Trypsin/0,53 mM EDTA in HBSS (Hank’s Balanced Salt Solution) with sodium bicarbonate, without calcium and magnesium.

  • Trypsin EDTA 1X 0,05% Trypsin/0,53 mM EDTA in HBSS with sodium bicarbonate, without calcium and magnesium
  • Porcine Parvovirus tested

Trypsin is used to enzymatically release adherent cells from tissue culture plates for passaging. Divalent cations such as calcium and magnesium, which are often present in the cell culture environment, inhibit this action. EDTA sequesters these ions and thus enhances thh efficacy of trypsin. This product contains trypsin at a concentration of 0,05% with 0,53 mM EDTA, and contains sodium bicarbonate.

Customers Who Bought This Also Bought