Order Entry
ContactUsLinkComponent
PBS 1X W/O CA+MG 6X500ML
Corning® PBS (Phosphate-Buffered Saline), Corning
Catalog #: CA45000-446
Supplier:  Corning
CAS Number:  
PBS 1X  W/O CA+MG 6X500ML
Corning® PBS (Phosphate-Buffered Saline), Corning
Catalog #: CA45000-446
Supplier:  Corning
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Description:
    Phosphate-Buffered Saline, 1X Without Calcium and Magnesium
  • Size:
    500 mL
  • Storage temperature:
    Ambient
  • Shipping temperature:
    Ambient
  • Storage Temperature:
    +2 °C…+30 °C
  • Supplier No.:
    21-040-CV

Specifications

About this item

Phosphate buffered saline (PBS) is a buffer solution containing sodium chloride, sodium phosphate, and in some formulations potassium chloride and potassium phosphate

  • Low endotoxin - < 0.25EU/mL
  • Mycoplasma tested
  • Liquid media products are formulated with cell culture grade WFI water
  • Manufactured in a cGMP environment

PBS helps to maintain a constant pH and is isotonic and non-toxic to cells.

Customers Who Bought This Also Bought