Order Entry
ContactUsLinkComponent
DULBECCOS PBS 1X 500ML PK6
Dulbecco's phosphate-buffered saline (DPBS), Mediatech
Catalog #: CA45000-430
Supplier:  Corning
CAS Number:  
Dulbecco's phosphate-buffered saline (DPBS), Mediatech
Catalog #: CA45000-430
Supplier:  Corning
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • ā€¢ State issued document with your organization's Federal Tax ID Number
  • ā€¢ State issued document with your organization's Resale Tax ID Number
  • ā€¢ City or County issued Business License
  • ā€¢ State Department of Health Services License
  • ā€¢ Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Description:
    DPBS, 1X with calcium and magnesium
  • Storage temperature:
    Ambient
  • Storage Temperature:
    +2 Ā°Cā€¦+30 Ā°C
  • Volume:
    500 ml
  • Supplier No.:
    21-030-CV

Specifications

About this item

Dulbecco's Phosphate Buffered Saline (DPBS) is a buffer solution which can be used to maintain cell culture media in the physiological pH range of 7.0 to 7.6

  • Low endotoxin - < 0.25EU/mL
  • Mycoplasma tested
  • Liquid media products are formulated with cell culture grade WFI water
  • Manufactured in a cGMP environment

Complete Certificate of Analysis available for each production lot, along with full formulation information.

It can serve as an irrigating, transporting or diluting fluid while maintaining cell tonicity and viability for a limited period of time.

Caution: Storage temperature: Solutions 2 - 30 Ā°C; Powder formats 2 - 8 Ā°C
Customers Who Bought This Also Bought