Order Entry
ContactUsLinkComponent
RPMI 1640 W/GLUTAMINE 500MLPK6
Corning® RPMI 1640, Corning
Catalog #: CA45000-396
Supplier:  Corning
CAS Number:  
Corning® RPMI 1640, Corning
Catalog #: CA45000-396
Supplier:  Corning
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Description:
    RPMI 1640 with L-glutamine
  • Storage temperature:
    Refrigerate
  • Media Format:
    Liquid
  • Cat. no.:
    CA45000-396
  • Supplier No.:
    10-040-CV

Specifications

About this item

Named for the site at which it was developed, the Roswell Park Memorial Institute, RPMI 1640 is a modification of McCoy's 5A containing an increased level of inositol.

  • Low endotoxin - < 0.25EU/mL
  • Mycoplasma tested
  • Liquid media products are formulated with cell culture grade WFI water
  • Manufactured in a cGMP environment

RPMI 1640 is also used for suspension cultures or monolayer human leukemic cells. Examples of cell types for which this medium is appropriate include mammalian myeloma and hybridoma cells such as mouse hybridomas, human leukocytes, and B and T lymphocytes.

Customers Who Bought This Also Bought