1860 Disposable N95 Health Care Particulate Respirator and Surgical Masks 3M™
Catalog #: CA43100-063
Supplier: 3M
Specifications
- Size:Regular
- Cat. no.:CA43100-063
- Supplier No.:1860
Specifications
About this item
This respirator has a soft inner shell for extended comfort on the face.
- Fluid-resistant to reduce the potential exposure of the wearer to blood and other body fluids
- Provides minimum filtration efficiency of 95% against certain solid and liquid particles that do not contain oil
Meets CDC guidelines for TB exposure control and helps reduce exposure to airborne particles.
Caution: All 3M™ products must be used in accordance with OSHA regulations and the user instructions, warnings, and limitations accompanying each product.
Customers Who Bought This Also Bought