WypAll® X60 Wipers
Catalog #: CA37002-024
Supplier: Kimberly-Clark
Specifications
- W×L:31.8×36.6 cm (12¹/₂×14³/₈")
- Color:White
- Packaging:Quarter-fold
- Supplier No.:34865
Specifications
About this item
WypAll® X60 cloths are absorbent, lightweight, and reinforced to stay strong in light-duty cleaning tasks.
- Embossed surface traps and holds dirt and liquids
- Can absorb more than 2× as much water as rags
- Can be used in a range of medium- to heavy-duty applications
- Contains no adhesives or binders, yet they hold together and don't leave residues behind
- Tough enough for big jobs, but soft enough for use on face and hands
- Can be used wet or dry
Durable, uniformly reinforced towels for extra scrubbing and cleaning power. Embossed surface traps and holds dirt and liquids. Can be used in a full range of applications including utility companies, plant and fleet maintenance, machine shops, ore mines, and grocery stores. No adhesives or binders; wipers leave nothing behind. Tough enough for big jobs, yet soft enough for face and hands. Available in a variety of sizes and formats. Wipers 21903-040 and 37002-024 are hygienic washcloths that are excellent for both body cleansing and drying.
Customers Who Bought This Also Bought