Order Entry
ContactUsLinkComponent
KIMTEX* Surface Preparation Wipers
KIMTECH PREP® KIMTEX™ Wipers, KIMBERLY-CLARK PROFESSIONAL®
Catalog #: CA37002-022
Supplier:  Kimberly-Clark
KIMTECH PREP® KIMTEX™ Wipers, KIMBERLY-CLARK PROFESSIONAL®
Catalog #: CA37002-022
Supplier:  Kimberly-Clark

Specifications

  • Color:
    Blue
  • Description:
    KIMTEX™ Wipers
  • L×W:
    31.7 x 33 cm (12.5 x 13")
  • Cat. no.:
    CA37002-022
  • Packaging:
    Quarter Fold
  • Supplier No.:
    33560

Specifications

About this item

KIMTECH PREP® KIMTEX™ Wipers feature 100% polypropylene construction for cloth-like feel and durability.

KIMTECH PREP® KIMTEX™ Wipers feature 100% polypropylene construction for cloth-like feel and durability.

Wipers provide a virtually lint-free cleaning performance and absorb quickly for big cleanups. Engineered for critical applications involving solvents, inks, and paints, they are also perfect for use with lubricants, oil, and grease. Acid-, base-, and solvent-resistant. Available in a variety of formats to meet every need. Color: blue. Applications include furniture finishing, engine assembly/repair, prepackaging wipe down, LCD panel assembly, appliance assembly, printing, and glass or window cleaning.

Customers Who Bought This Also Bought