Nunclon™ Delta 96-Well MicroWell™ Optical Bottom Plates, Sterile, Thermo Scientific
Catalog #: CA37000-558
Supplier: Thermo Fisher Scientific
Specifications
- Well Volume:200 µL
- Color:Black
- Growth Area:0.32 cm2
- Working Volume:200 µl
- Supplier No.:165305
Specifications
About this item
These polystyrene plates provide optimum clarity for viewing well contents
The Nunclon™Δ treated surface ensures optimal cell growth and attachment. Plates feature either a black or white polystyrene upper structure and a polymer bottom. The flat-bottom well design facilitates plate reader access. The 96-well footprint is compatible with standard equipment and automated systems.
Ordering information: Both plates come supplied with lids. For sealing tapes, see the 62409-118 series.
Customers Who Bought This Also Bought