Filter Forceps, Cytiva (Formerly Pall Lab)
Catalog #: CA34181-102
Supplier: Cytiva
Specifications
- Description:Forceps, Multi-Colored
- Cat. no.:CA34181-102
- Supplier No.:4690
Specifications
About this item
For easy placement of membrane filters into filter funnels or petri dishes.
- Flat surface
- Avoid membrane damage
- Stainless steel
Tips have a flat, smooth surface to avoid membrane filter damage. Stainless steel construction with polypropylene finger grips for a comfortable and secure hold. Autoclavable.
Customers Who Bought This Also Bought