ContactUsLinkComponent
Goggle
Uvex Flex Seal® Safety Goggles, Honeywell Safety
Catalog #: CA33002-548
Goggle
Uvex Flex Seal® Safety Goggles, Honeywell Safety
Catalog #: CA33002-548
Specifications
  • Headband:
    Fabric
  • Supplier No.:
    S3405X
Specifications
About this item
Lightweight silicone body conforms to the face to protect against impacts and chemical splashes. The silicone is cured to stay soft and supple even in extreme temperatures.
Lightweight silicone body conforms to the face to protect against impacts and chemical splashes. The silicone is cured to stay soft and supple even in extreme temperatures. Low-profile and optically correct clear polycarbonate lens provides the full field of vision. Uvextreme® antifog lens coating and indirect venting system minimize fogging. Goggles fit over most prescription eyewear and feature pivoting clips that permit the wearer to position them on a hard hat or position the headband above the ears. Frame color: navy.
  • Provide excellent impact and chemical-splash resistance
  • Fit over most prescription eyewear
Low-profile and optically correct clear polycarbonate lens provides the full field of vision. Uvextreme® antifog lens coating and indirect venting system minimize fogging. Goggles fit over most prescription eyewear and feature pivoting clips that permit the wearer to position them on a hard hat or position the headband above the ears. Frame color is navy.
Certifications: Goggles meet ANSI Z87+ and CSA Z94.3 standards.
Customers Who Bought This Also Bought