Centurion™ 454 Series Splash Safety Goggles, 3M™ Company
Catalog #: CA33001-603
Supplier: 3M
Specifications
- Lens Coating:Anti-fog
- Description:Centurion™ 454 series safety goggles with splash protection and adjustable headband
- Supplier No.:40305-00000-10
Specifications
About this item
Centurion™ safety splash goggle is a reliable, economical choice in protective eyewear. Stable, sturdy construction will protect wearer's eyes.
- Wide-angle wraparound lens and frame for good peripheral vision
- Indirect venting system to help minimize fogging
- Accommodates most prescription eyewear
Centurion™ safety splash goggle is a reliable, economical choice in protective eyewear. Stable, sturdy construction will protect wearer's eyes.
Includes wide nasal flare for added comfort and an indirect venting system to minimize fogging. All goggles feature a strap attachment system.
Certifications: Meets ANSI Z87.1-2010, CSA Z94.3-2007.
Customers Who Bought This Also Bought