Uvex Astro OTG™ 3001 Safety Eyewear, Honeywell Safety
Catalog #: CA33000-517
Supplier: Honeywell Safety Products
Specifications
- Lens Coating:Scratch-resistant/hardcoat (Ultra-dura®)
- Lens Tint:Clear
- Frame Color:Blue
- Supplier No.:S2510
Specifications
About this item
Specially designed to fit comfortably and attractively over prescription eyewear.
Meets ANSI Z87+ and CSA Z94.3 standards.
- Lens inclination provides a close-to-the-face fit
- Adjustable Duoflex™ temples feature comfort cushions for extended use
Specially designed to fit comfortably and attractively over prescription eyewear.
Patriot® RWB frames have a red, white, and blue design with miniature U.S. flags on the temples. Easily replaceable lenses are available with scratch- and chemical-resistant Ultra-dura® coating or Uvextreme® AF antifog, antistatic, anti-scratch, and anti-UV coating.
Certifications: Meets ANSI Z87+ and CSA Z94.3 standards.
Customers Who Bought This Also Bought