TouchNTuff® 92-500 Nitrile Gloves, Powdered, Ansell
Catalog #: CA32490-002
Supplier: Ansell Healthcare
Specifications
- Ambidextrous/Hand-specific:Ambidextrous
- Glove Color:Green
- Glove size:L
- Length:24 cm (9 3/8")
- Powdered/Powder-free:Powdered
- Disposable / reusable:Disposable
- Latex/Latex-free:Latex-free
- Thickness:4.7 mil
- Cuff style:Rolled, beaded
- Finger thickness:5.6 mil
- Palm thickness:4.7 mil
- Cuff thickness:4 mil
- Tensile strength:14 MPa (before aging), 14 MPa (after aging)
- Ultimate elongation:500% (before aging), 400% (after aging)
- Finish:Textured
- Cat. no.:CA32490-002
- Supplier No.:9250011090
Specifications
About this item
Made of 100% nitrile, TouchNTuff® 92-500 disposable gloves are thin yet strong, providing both tactile sensitivity and splash resistance against hazardous chemicals.
- Excellent fingertip sensitivity
- Exceptional durability
- Lightly powdered and lightweight
- Silicone-free
They offer three times the puncture resistance of standard latex or vinyl gloves. These gloves combine excellent fingertip sensitivity with exceptional durability to give a high level of protection against grease and oil. They are lightly powdered, lightweight and comfortable to wear.
Certifications: TouchNTuff® gloves comply with FDA food handling requirements (21 CFR 177.2600).
Packaging: Gloves are packaged 100 gloves in a box; 10 boxes in a carton.
Customers Who Bought This Also Bought