Whatman™ Sealing Film and Foil for Microplates, Whatman products (Cytiva)
Catalog #: CA28317-478
Supplier: Cytiva
Specifications
- Color:Clear
- Description:Thin Cold Sealing Film with Adhesive Backing
- Material:Polyester
- Thickness:50 µm
- Cat. no.:CA28317-478
- Supplier No.:7704-0001
Specifications
About this item
Seals are used to control humidity and reduce evaporation of samples. They prevent spills and contamination.
- Cold seals in clear polyester or PP are self-sticking with inert adhesive
- Heat seals are suitable for PP microplates only and are applied using heat and pressure
Customers Who Bought This Also Bought