ContactUsLinkComponent
Protein Saver Card
Whatman™ 903 Proteinsaver Sample Collection Card, Whatman products (Cytiva)
Catalog #: CA28170-017
Supplier:  Cytiva
Protein Saver Card
Whatman™ 903 Proteinsaver Sample Collection Card, Whatman products (Cytiva)
Catalog #: CA28170-017
Supplier:  Cytiva
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    903 Protein Saver Card (US)
  • Cat. no.:
    CA28170-017
  • Supplier No.:
    10534612
Specifications
About this item
Whatman 903 sample collection cards are designed for the collection of biological fluids
  • Ready to use cards save time and costs during sample collection, transport, and archiving
  • Strict quality control and GMP manufacturing standards ensure high quality, reproducibility, and purity
  • Choice of five-spot Proteinsaver card or four-spot Proteinsaver snap-apart card
The sample collection area of the Whatman™ 903 Proteinsaver card contains five half-inch circles. Each circle holds 75 to 80 µl of sample. Wraparound cover has spaces for name and date of collection and is imprinted with the universal biohazard symbol. The Proteinsaver card fits into Whatman foil barrier resealable Ziploc® bags for storage.
The cards are ready to use and provide easy collection, identification, and transport of samples, without the need for costly freezers or dry ice delivery.
Certifications: Under current regulations, specimen collection cards, including neonatal screening cards, which are to be used for human diagnostic tests, are classified as "other" IVD devices under the European IVD Directive and require the CE mark if sold within the European Union. All 903 specimen collection devices manufactured and printed by Whatman for neonatal testing in the EU undergo post-printing quality control and carry the CE mark. Whatman will assume no responsibility for the quality or performance of 903 collection devices converted, printed, or packaged by third party suppliers.
Caution: Protein saver cards are for research use only. Whatman will assume no responsibility for the quality or performance of 903 collection devices converted, printed, or packaged by third party suppliers.
Customers Who Bought This Also Bought