Order Entry
ContactUsLinkComponent
MF-Endo Broth (Plastic Ampule)
Microbiological Media in Ampoules, Cytiva (Formerly Pall Lab)
Catalog #: CA28150-500
Supplier:  Cytiva
CAS Number:  
MF-Endo Broth (Plastic Ampule)
Microbiological Media in Ampoules, Cytiva (Formerly Pall Lab)
Catalog #: CA28150-500
Supplier:  Cytiva
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Packaging:
    2 mL Plastic Ampule
  • Description:
    MF-Endo Broth, Total Coliforms
  • Cat. no.:
    CA28150-500
  • Supplier No.:
    68105

Specifications

About this item

A wide variety of sterile ampoule media for common microbiological analyses, in clear glass or plastic ampoules. Supplied pre-measured for maximum efficiency and convenience. For public wastewater and drinking water: MF-Endo and M-FC (with and without rosolic acid for helping to increase the specificity of the medium for faecal coliforms) media are used when analysing water samples for total coliforms, faecal coliforms and E. coli. For food and beverage: M-TGE, HPC, M-Green YM, and orange serum broths are used in the food and beverage industry to QC final products (and fluids used in the manufacture of these products) to detect potential spoilage organisms. Trypticase soy broth, M-TGE, and HPC may be used to test final products and process waters for total bacteria in the pharmaceutical and microelectronics industries. Pseudomonas and KF-Streptococcal broths are highly selective media for organisms in these genera, and are used to monitor samples wherever contamination by these organisms must be prevented.

  • 2 ml ampoules are a practical, time saving aid for culturing microorganisms on membrane filters
  • Ready to use, no dilutions necessary
  • Shelf life up to 1 year storing at 2 to 8 °C
  • Meet Membrane Filter (MF) Technique Standard Methods requirements with 2 ml ampoules (requirements referenced in Standard Methods for the Examination of Water and Wastewater, 1992 APHA, AWWA, WEF, 20th Edition)
Customers Who Bought This Also Bought