Order Entry
ContactUsLinkComponent
TCLP Glass Fiber Filter
TCLP Glass Fiber Filters, Cytiva (Formerly Pall Lab)
Catalog #: CA28149-455
Supplier:  Cytiva
TCLP Glass Fiber Filter
TCLP Glass Fiber Filters, Cytiva (Formerly Pall Lab)
Catalog #: CA28149-455
Supplier:  Cytiva

Specifications

  • Ø:
    4.7 cm
  • Particle retention:
    0.7 µm
  • Porosity:
    Fine
  • Flow rate:
    Medium
  • Cat. no.:
    CA28149-455
  • Supplier No.:
    66258

Specifications

About this item

TCLP glass fiber filters are designed to meet requirements for use in U.S. EPA SW-846 Method 1311: Toxic Characteristics Leaching Procedure (TCLP).

  • Reduces filtration costs and premature clogging when filtering difficult-to-filter or highly contaminated solutions
  • Extends filter life and reduces final filter changes with high capacity prefilters
  • Eliminates sample contamination, binder-free borosilicate glass fiber has no added extractables

Filters a wide range of particulate loads and viscous solutions with a selection of filter thicknesses from which to choose.

Customers Who Bought This Also Bought