Order Entry
ContactUsLinkComponent
GN-6 Metricel* Membrane Disc Filter
GN Metricel® Membrane Disc Filters, Cytiva (Formerly Pall Lab)
Catalog #: CA28148-642
Supplier:  Cytiva
GN-6 Metricel* Membrane Disc Filter
GN Metricel® Membrane Disc Filters, Cytiva (Formerly Pall Lab)
Catalog #: CA28148-642
Supplier:  Cytiva

Specifications

  • Ø:
    47 mm
  • Pore size:
    0.45 µm
  • Sterility:
    Non sterile
  • Description:
    GN-6 Metricel Filter, 0.45 µm, Gridded
  • Packaging:
    Bulk
  • Type:
    Grid
  • Supplier No.:
    63020
  • Gamma-Irradiated:
    No

Specifications

About this item

The GN-6 membrane is certified for the microbiological analysis of potable, waste, process, and natural waters in accordance with the Membrane Filter Technique referenced in Standard Methods for the Examination of Water and Wastewater.

GN-6 and GN-4 Metricel® membrane filters are made of mixed cellulose esters. GN-6 grids have a unique dot grid pattern that provides easy quantification of bacterial colonies without growth inhibition or enhancement. GN-4 membranes dissolve completely using standard digestion procedures, and clear completely for minimal interference in fiber counting.

  • For microbiological analysis
  • Mixed cellulose ethers
  • Easy bacterial quantification

The GN-6 membrane is certified for the microbiological analysis of potable, waste, process, and natural waters in accordance with the Membrane Filter Technique referenced in Standard Methods for the Examination of Water and Wastewater.

Ideal for isolation and enumeration of total and fecal coliforms, E.

GN-6 and GN-4 Metricel® membrane filters are made of mixed cellulose esters. GN-6 grids have a unique dot grid pattern that provides easy quantification of bacterial colonies without growth inhibition or enhancement. GN-4 membranes dissolve completely using standard digestion procedures, and clear completely for minimal interference in fiber counting.

E. coli, fecal Streptococcus, fungi, and other heterotrophic bacteria. The GN-4 membrane is suitable for air monitoring applications and meets NIOSH requirements for airborne metals and asbestos monitoring.

Certifications: Meets NIOSH requirements for airborne metals and asbestos monitoring.
Ordering information: GN-6 is available gamma irradiated, individually packaged (except filter model 60016) or nonsterile, bulk packed. GN-4 is available in nonsterile bulk packs or cavities, with or without support pads.
Customers Who Bought This Also Bought