To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Filter Media:Glass Fiber/Bio-Inert® Nylon
- Pore size:3.0 µm/ 0.2 µm
- Well Volume:1 ml
- Cat. no.:CA28148-626
- Working Volume:900 µl
- Supplier No.:5053
Specifications
About this item
Constructed of chemically resistant, biologically inert polypropylene, plates are optimized for a variety of applications requiring larger per well volume processing.
- Chemically resistant material
- Extended flow directors
- Individually sealed wells
Plates feature extended flow directors to eliminate splattering and crosstalk of filtrate, a serialized bar code label for use in automated tracking systems, robotics-friendly design, standard SBS format, and individually sealed wells to eliminate crosstalk.
Available in a variety of membranes and membrane configurations to meet various application needs. Choose from GHP (hydrophilic polypropylene), glass fiber (borosilicate glass without binder), glass fiber over Bio-Inert® modified low protein-binding nylon, hydrophobic PTFE, Supor® polyethersulfone, or Omega™ polyethersulfone (PES) membranes.