Specifications
- Pore size:0.8/0.2 µm
- Description:AcroPak™ 500
- Cat. no.:CA28146-177
- Connection:Stepped hose barb
- Effective Filtration Area:500 cm² (78 in²)
- Supplier No.:12991
Specifications
About this item
A filter for every step in your process. From drug discovery and basic research to process development and production, Pall Laboratory’s AcroPak capsules come in a variety of membrane types and volume sizes.
- 100% integrity tested (where noted)
- Endotoxin level: <0.25 EU/ml using Limulus Amebocyte Lysate test
- Bacterial retention: 0.2 μm lot samples retain 10⁷ cfu/cm² of B. diminuta per modified ASTM F838
- Mycoplasma reduction: 0.1 μm lot samples retain 10⁷ cfu/cm² A. Laidlawii per modified ASTM F838
- Biological safety: Passes United States Pharmacopeia (USP) biological reactivity test in vivo <88>
- Sterilisation by gamma irradiation
Primary applications include: Buffer media and reagent manufacturing, protein purification and cell culture and bioburden filtration.
Supor PES (Polyethersulfone):
With high flow and capacity, broad fluid compatibility, low protein adsorption, active ingredients, and stabilizers, Pall PES membranes are best suited for filtration of preparative fluids such as buffers, media, and bioprocess intermediates.
Supor EKV Asymmetric PES:
PES filters with asymmetric membrane layers have the same benefits of Supor PES (above), but with additional debris holding. This makes them ideal for high turbidity, high bioburden solutions. All Supor EKV products are a 0.65 µm pre-filter and a 0.2 µm asymmetric membrane.
Fluorodyne II PVDF (hydrophilic polyvinylidene fluoride):
Low extractables, low product and excipient adsorption, simple validation for the sterilization of a wide range of formulations make PVDF membranes ideal for final filtration.