Order Entry
ContactUsLinkComponent
Mini Capsule Filter, 0.45um, Sterile
Mini Capsule Filter, 0.45 µm, Sterile, Cytiva (Formerly Pall Lab)
Catalog #: CA28145-112
Supplier:  Cytiva
Mini Capsule Filter, 0.45 µm, Sterile, Cytiva (Formerly Pall Lab)
Catalog #: CA28145-112
Supplier:  Cytiva

Specifications

  • Cat. no.:
    CA28145-112
  • Maximum Operating Pressure:
    3.4 bar (50 psid)
  • Effective Filtration Area:
    500 cm2
  • Maximum Operating Temperature:
    88 °C (190 °F)
  • Supplier No.:
    12123

Specifications

About this item

Use both for prefiltration of particulate liquids prior to final sterilizing filtration, and for removal of particles and debris from liquids not requiring final sterilization.

  • Two layers
  • Barbed hose fittings
  • Polycarbonate housing

Incorporates two layers of Versapor® (acrylic copolymer) membrane: 0.8µm prefilter layer followed by 0.45 µm final filter.

Polycarbonate housing is fitted with 12.7 mm (¹/₂") male, barbed, hose fittings for rapid installation with flexible tubing. Biosafe and disposable. Gamma irradiation sterilized.

Not autoclavable and not suited for dry-heat applications.

Customers Who Bought This Also Bought