Order Entry
ContactUsLinkComponent
Easy-Pressure Syringe Filter Holder, 25mm
Easy-Pressure Syringe Filter Holder, 25 mm, Cytiva (Formerly Pall Lab)
Catalog #: CA28144-109
Supplier:  Cytiva
Easy-Pressure Syringe Filter Holder, 25mm
Easy-Pressure Syringe Filter Holder, 25 mm, Cytiva (Formerly Pall Lab)
Catalog #: CA28144-109
Supplier:  Cytiva

Specifications

  • Filter Diameter:
    3.5 mm (1⅜")
  • Length:
    2.7 cm (11/16")
  • Cat. no.:
    CA28144-109
  • Supplier No.:
    4320

Specifications

About this item

Made of Delrin® plastic with Type 316 stainless steel membrane support screen.

  • Inlet and outlet seal plugs included
  • Autoclavable
  • Viton® rubber O-ring

Accepts 25 mm Ø membrane filters (recommended prefilter size: 20 mm). Fits any standard syringe with Luer-type fitting.

Customers Who Bought This Also Bought