Order Entry
ContactUsLinkComponent
Acrodisc* Premium Syringe Filter
Acrodisc® Syringe Filters for Prefiltration, Cytiva (Formerly Pall Lab)
Catalog #: CA28143-986
Supplier:  Cytiva
Acrodisc* Premium Syringe Filter
Acrodisc® Syringe Filters for Prefiltration, Cytiva (Formerly Pall Lab)
Catalog #: CA28143-986
Supplier:  Cytiva

Specifications

  • Ø:
    25 mm
  • Pore size:
    1.00 µm
  • Sterility:
    Non sterile
  • Membrane:
    Glass Fiber
  • Supplier No.:
    4523

Specifications

About this item

For prefiltering heavily contaminated aqueous or solvent based samples.

  • Increased pore size for reduced clogging
  • Maximized throughput
  • Wide range of pore sizes available

For prefiltering heavily contaminated aqueous or solvent based samples.

The glass fiber membrane has very low extractables. Filters are made of 100% binder-free borosilicate glass fiber.

The Versapor® (acrylic copolymer) membrane filters feature large pore sizes ideal for prefiltration. These filters are color-coded by pore size for easy identification.

Polypropylene housing, female Luer-lock inlet and male slip Luer outlet.

Certifications: Acrodisc PSF syringe filters are automation certified to assure smooth operation and worry-free performance 24 hours a day in automated workstations.
Packaging: Syringe filters packed in cases of 200 are available in four packs of 50 or in an AutoPack™ format. The AutoPack™ format consists of eight tubes with 25 syringe filters stacked on top of each other, which is specifically designed for the Caliper™ robotic workstations.
Customers Who Bought This Also Bought