Order Entry
ContactUsLinkComponent
Refill Dye
Tissue Marking Dyes (TMD™), TBS®, General Data
Catalog #: CA27900-262
CAS Number:  
Tissue Marking Dyes (TMD™), TBS®, General Data
Catalog #: CA27900-262
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Color:
    Yellow
  • Description:
    Tissue Marking Dyes (TMD™)
  • Size:
    8 oz
  • Cat. no.:
    CA27900-262
  • Supplier No.:
    TMD-Y

Specifications

About this item

Ideal for marking margins or for processing multiple specimens in one tissue cassette, these dyes have been reformulated for superior adherence to tissue surfaces

Viscosity allows a thin, even coating of pigment without masking underlying tissue. Quick drying decreases frozen sectioning time and provides excellent readability. Size: 237 mL (8 oz.).

Customers Who Bought This Also Bought