ContactUsLinkComponent
PDLE TSTR TTL BAC/YST/MD 10/PK
Paddle Tester, Total Aerobic Bacteria/Yeast and Mold, Hach
Catalog #: CA26108-10
Supplier:  Hach
CAS Number:  
Paddle Tester, Total Aerobic Bacteria/Yeast and Mold, Hach
Catalog #: CA26108-10
Supplier:  Hach
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
  • Cat. no.:
    CA26108-10
  • Supplier No.:
    26108-10
Specifications
About this item
Hach Paddle Testers allow fast, easy screening of water samples, solid surfaces, and nonviscous liquids where unsanitary conditions may exist
The paddle is a double-sided slide attached to the vial cap. Each side of the slide is used to perform a separate test. The slide has a molded-in grid for easy colony counting without opening the optically clear, leakproof vial. Paddle Tester, Total Aerobic Bacteria/Yeast and Mold (Tryptone Glucose Extract Agar/Sabouraud Dextrose Agar), pk/10.
Certifications: UL Listed.
Ordering information: Accessories not included.
Customers Who Bought This Also Bought